Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STK11IP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111369
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111369 Supplier Invitrogen™ Supplier No. PA5111369
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVLEPDAHAA VQELLAVLTP VTNVAREQLG EARDLLLGRF QCLRCGHEFK PEEPRMGLDS EEGWRPLFQK TESPAVC Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 66%.

May regulate STK11/LKB1 function by controlling its subcellular localization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK11IP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STK11IP
Gene Accession No. Q8N1F8
Gene Alias 1200014D22Rik; BB131189; KIAA1898; LIP1; LKB1-interacting protein 1; LKB1IP; serine/threonine kinase 11 interacting protein; serine/threonine kinase 11-interacting protein; serine/threonine-protein kinase 11-interacting protein; STK11 interacting protein; STK11IP; STK11IP1
Gene Symbols STK11IP
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human STK11IP. Recombinant protein control fragment (Product #RP-106445).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114790
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.