Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STK16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529415
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529415 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529415 Supplier Invitrogen™ Supplier No. PA529415
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (93%), Rat (95%), Rhesus Monkey (98%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

STK16 (PKL12) is a serine/threonine kinase that functions in signal transduction pathways and transcriptional regulation. Protein kinase that act on both serine and threonine residues. STK16 may regulate transforming growth factor-beta (Tgfb) and vascular endothelial growth factor transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene STK16
Gene Accession No. O75716
Gene Alias Edpk; Embryo-derived protein kinase; F52; hPSK; kinase related to cerevisiae and thaliana; Krct; MPSK; Mpsk1; Myristoylated and palmitoylated serine/threonine-protein kinase; PKL12; protein kinase expressed in day 12 fetal liver; Protein kinase Krct; Protein kinase PKL12; serine/threonine kinase 16; serine/threonine-protein kinase 16; Stk16; TGF-beta-stimulated factor 1; Tsf1; TSF-1; tyrosine-protein kinase STK16
Gene Symbols STK16
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human STK16. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8576
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.