Learn More
Eurofins Discoverx STK25, active, 250 µg. (14-929M)
Supplier:  Eurofins Discoverx 14929M
Encompass_Preferred
N-terminal GST-tagged and C-terminal 6xHis-tagged recombinant, human STK25 amino acids 1-308. STK25 is a member of the germinal centre kinase III (GCK III) subfamily of the sterile 20 superfamily of kinases. The enzyme plays a role in serine-threonine liver kinase B1 (LKB1) signaling pathway to regulate neuronal polarization and morphology of the Golgi apparatus. The protein is translocated from the Golgi apparatus to the nucleus in response to chemical anoxia and plays a role in regulation of cell death. A pseudogene associated with this gene is located on chromosome 18. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9432
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.