Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STK40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526629
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526629 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526629 Supplier Invitrogen™ Supplier No. PA526629
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STK40 Polyclonal Antibody for Western Blot, IHC (P), Flow

STK40 is a protein coding gene. May be a negative regulator of NF-kappa-B and p53-mediated gene transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK40
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene STK40
Gene Accession No. Q7TNL3, Q8N2I9
Gene Alias 2310004N11Rik; AI428792; Lyk4; ortholog of Ser/Thr-like protein kinase Lyk4; RGD1307310; RP11-268J15.4; Ser/Thr-like kinase; Ser/Thr-like protein kinase lyk4; serine/threonine kinase 40; serine/threonine-protein kinase 40; Serine/threonine-protein kinase lyk4; SgK495; SHIK; SINK-homologous serine/threonine-protein kinase; Stk40; Sugen kinase 495
Gene Symbols STK40
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 372-403 amino acids from the C-terminal region of human STK40.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74178, 83931
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.