Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STMN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 720178 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-017-8 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-017-8 Supplier Invitrogen™ Supplier No. 720178
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature.

STMN2, also known as SCG-10, is a neuronal growth-associated protein that shares significant amino acid sequence similarity with the phosphoprotein stathmin. STMN2 may play a role in neuronal differentiation, and in modulating membrane interaction with the cytoskeleton during neurite outgrowth. Reductions in the expression of this gene have been associated with Down's syndrome and Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STMN2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2-7.4
Gene Stmn2
Gene Accession No. P21818, P55821, Q93045
Gene Alias AI159727; hypothetical protein LOC534991; neuronal growth-associated protein (silencer element); neuron-specific growth-associated protein; Protein SCG10; SCG10; Scgn10; SGC10; stathmin 2; stathmin-2; stathmin-like 2; Stmb2; Stmn2; superior cervical ganglia, neural specific 10; superior cervical ganglion-10 protein; superiorcervical ganglia, neural specific 10
Gene Symbols Stmn2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STMN2 (aa 29-179).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11075, 20257, 84510
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.