Learn More
Enzo Life Sciences STO-609 (25mg). CAS: 1173022-21-3
SDP
Supplier:  Enzo Life Sciences BMLEI3890025
Encompass_Preferred
STO-609 is a specific CaM kinase kinase (CaM-KK) inhibitor. Ki=0.2μM and 0.04μM for α and β isotypes respectively. Demonstrates >80-fold selectivity for CAM-KK over CaMK1, CaMK2, CaMK4, MLCK, PKA, PKC, and p42 MAPK. Concentrations up to 10μg/ml do not affect cellular viability. It is a tool for exploring the role of the CaM-KK pathway in various physiological processes. Alternative name: 7-Oxo-7H-benzimidazo[2,1-a]benz[de]isoquinoline-3-carboxylic acid, acetate salt. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0766
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.