Learn More
Med Vet International Stockinette 3" X 25 Yds.
Supplier:  Med Vet International SN3
Encompass_Preferred
Heavyweight, premium-grade, tubular stockinette for use under and over casts. Protects skin under plaster casts. Dispenser box simplifies cutting for exact fit. Can also be used for other dressing applications. 25 yards per box.
Catalog No. 50-283-0741