Learn More
Poltex Stockinette Dispenser for Countertop
SDP
Catalog No. 21377568
Change view
Click to view available options
Includes:
None
Catalog No. Includes
21-377-568 None
1 options
Catalog No. 21-377-568 Supplier Poltex Supplier No. STCKDISPCT
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Stockinette dispenser for countertop or cart top placement.
  • Stockinette dispenser for countertop or cart top placement
  • Fits multiple sizes
  • Front slant justifies rolls toward front
  • Two different slots to dispense from depending on roll size
  • Rubber bumpons on base for non-slip placement
  • Durable, cleanable PETG construction
  • Optional side bracket for scissors
Specifications
Type Dispensing Bin
Material PETG
Color Clear
Shape Rectangular
Mounting Type Freestanding
Autoclavable Not Autoclavable
Holds Stockinette Rolls
Includes None
Style Hinged Lid with Front Opening