Learn More
Poltex Stockinette Dispenser with Wall Mount
SDP
Catalog No. p-200089259
Change view
Click to view available options
Includes:
None
Side Bracket
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-377-570 None
21-377-571 Side Bracket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-377-570 Supplier Poltex Supplier No. STCKDISPW
Only null left
Add to Cart
Add to Cart
Stockinette dispenser with screws and anchors to mount on a wall.
  • Stockinette dispenser for countertop or cart top placement
  • Fits multiple sizes
  • Front slant justifies rolls toward front
  • Two different slots to dispense from depending on roll size
  • Durable, cleanable PETG construction
  • Optional side bracket for scissors
  • Screws and anchors included for mounting
Specifications
Type Dispensing Bin
Material PETG
Color Clear
Shape Rectangular
Mounting Type Wall Mount
Autoclavable Not Autoclavable
Holds Stockinette Rolls
Includes None
Style Hinged Lid with Front Opening