Learn More
Stoelting™ Stoelting Rodent Warmer X2
SDP
Catalog No. 10000592 Shop All Stoelting Co Products
Change view
Click to view available options
Includes:
2 cage heating pads
2 mouse heating pads
2 rat heating pads
Cage heating pad
Mouse and cage heating pads
Mouse heating pad
Rat and cage heating pads
Rat and mouse heating pads
Rat heating pad
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
10-000-592 Rat and mouse heating pads
10-000-585 Cage heating pad
10-000-586 2 cage heating pads
10-000-587 Mouse heating pad
10-000-588 Mouse and cage heating pads
10-000-589 2 mouse heating pads
10-000-590 Rat heating pad
10-000-591 Rat and cage heating pads
10-000-593 2 rat heating pads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 10-000-592 Supplier Stoelting™ Supplier No. 53850RM
Add to Cart
Add to Cart
Stoelting™ Rodent Warmer X2 is for use before, during and after surgical procedures to improve surgical outcome and overall longevity.
  • Warming an animal the minimizes heat loss and improve surgical outcomes
  • Integrated touch screen allows for easy programming and controls 2 independent heating pads simultaneously
  • Available with a combination of mouse, rat or home-cage heating pads
  • Rectal Thermal probe sold separately
Control two independent heating pads simultaneouslyMinimize heat loss and improve surgical outcomes with Stoelting's Rodent Warmer X2.Features:Integrated touch screen for easy programming Controls two independent heating pads simultaneouslyRectal probe (sold separately) for body temperature feedbackAvailable with a combination of mouse, rat or cage size heating pads Heating PadsStoelting's Rodent Warmer can be used as a general warming system or with a rectal probe (sold separately) for more accurate, core temperature monitoring during pre- and post-op surgical procedures.Pre-Op: Heating pad can be placed underneath an induction chamber to reduce heat loss during anesthetic administration. During: Place heating pad on a rodent surgery table or stereotaxic instrument to maintain and monitor temperature during surgical procedures. Mouse heating pad designed for use with Stoelting's Just for Mouse (51730) and Just for Mice (51725) Stereotaxic Instrument or Mouse and Neonatal Rat Adaptor (51625). Rat heating pad designed for use with Stoelting's Lab Standard (51600) or New Lab Standard (51500) Stereotaxic Instrument.Post-Op: Cage heating pads can be placed in the animal's home cage for faster recovery following surgical procedures.
Specifications
For Use With (Application) Neuroscience
Includes Rat and mouse heating pads