Learn More
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications
Specifications
| Cross Reactivity | Human |
| Type | Stomach Antrum Tissue Slides (Normal) |
| For Use With (Application) | Immunohistochemistry-Paraffin |
| Regulatory Status | This product is for research use only and is not approved for use in humans or in clinical diagnosis. |
| Species | Human |
| Tissue Condition | Normal |
| Product Line | Novus |
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
