Learn More
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications
Specifications
| Cross Reactivity | Human |
| Type | Stomach Tissue Slides (Mucinous Adenocarcinoma) |
| Regulatory Status | This product is for research use only and is not approved for use in humans or in clinical diagnosis. |
| Species | Human |
| Tissue Condition | Mucinous Adenocarcinoma |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
