Learn More
PIP™ Stone™ Safety Goggles
Catalog No. 19036880 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant, Impact-resistant, UV-treated
Catalog No. Lens Coating
19-036-880 Anti-fog, Scratch-resistant, Impact-resistant, UV-treated
1 options
This item is not returnable. View return policy
Catalog No. 19-036-880 Supplier PIP™ Supplier No. 251600020
  • $145.10
    $106.65 / Pack of 5
    Save $38.45
    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,515.00
    $1,113.65 / Case of 12 PK
    Save $401.35
    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Helps provide protection from projectiles and impact in general use applications.
  • Clear polycarbonate lens with anti-scratch/anti-fog coating with high-impact protection
  • Adjustable elastic strap provides a comfortable fit
  • Indirect lower vents
  • Optional ANSI rated polycarbonate visor
  • can be attached and removed in seconds
  • 5 base curve lens
Compliance
Meets ANSI Z87.1-2015 and ANSI D3 standards.
Polycarbonate lenses block 99.99% of the sun’s ultraviolet rays. This ultraviolet protection has already been incorporated within the lens material when the lenses are being produced.
Specifications
Lens Coating Anti-fog, Scratch-resistant, Impact-resistant, UV-treated
Certifications/Compliance ANSI Z87.1-2015, ANSI D3
Description 5 base curve
Lens Tint Clear
Product Type Safety Goggles
For Use With (Application) Protection from projectiles and impact
Frame Color Transparent Gray
Headband Material Elastic
Size Universal
Ventilation Details Indirect
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material PVC
UV Protection UV400
Frame Type Indirect Vented