Learn More
Hach Company Stopper, Neoprene, One Hole, Size 8, 1/pk
Supplier:  Hach Company 1480477
Encompass_Preferred
Made of synthetic rubber that resists petroleum products, fats, oils, and organic chemicals. Recommended where natural rubber would deteriorate or be attacked.
Catalog No. 50-199-5121
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.