Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Snap-on Stopper



Provide airtight covers
$390.50 - $591.00
Specifications
Autoclavable | Autoclavable |
---|---|
Product Type | Plug Stopper |
For Use With (Equipment) | Serum bottles and vials |
Catalog Number | Mfr. No. | Color | Material | Diameter (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Material | Diameter (Metric) | Price | Quantity & Availability | |||
06-447C
![]() |
DWK Life Sciences
224100080 |
Red | Rubber | 13 mm |
Case of 1000 for $390.50
|
|
|||
06-447F
![]() ![]() |
DWK Life Sciences
W224100181 |
Gray | Chlorobutyl Rubber | 20 mm |
Case of 1000 for $581.50
|
![]() |
|||
06-447K
![]() |
DWK Life Sciences
224100180 |
Red | Rubber | 20 mm |
Case of 1000 for $591.00
|
|
|||
Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
Autoclavable | |
Plug Stopper | |
Serum bottles and vials |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title