Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Storage Flask
GREENER_CHOICESDP

Catalog No. 31501822 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
31-501-822 500 mL
31-502-331 250 mL
31-501-823 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-822 Supplier Synthware™ Supplier No. S520500
Add to Cart
Add to Cart

Storage Flask features a #15 O-ring joint on the sidearm and a high vacuum valve to control gas and/or liquid flow into the vessel.

  • High vacuum valve controls gas or liquid flow into the vessel
  • #15 O-ring joint on the side arm
  • Includes one #16 Viton™ O-ring

Specifications

Color Clear
Capacity (Metric) 500 mL
Material Borosilicate Glass
For Use With (Application) Organic Synthesis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.