Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Storage Plate

Catalog No. 07001131 Shop All Greiner Bio-One Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
No. per Pack:
1 Ea.
5/Pk.
50/Pk.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility No. per Pack
07-001-131 Non-sterile 1 Ea.
07-001-130 Sterile 1 Ea.
07-001-129 Non-sterile 50/Pk.
07-001-128 Non-sterile 5/Pk.
07-001-127 Sterile 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07-001-131 Supplier Greiner Bio-One Supplier No. 780270
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for applications requiring enhanced chemical and thermal resistance, MASTERBLOCK plates feature an ANSI-standard footprint for automation and are designed to facilitate sample handling and retrieval.

  • Suitable for sample storage, reagent housing, chromatography fraction collection, bacteria, yeast and cell cultures.
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
  • Sterile where indicated
  • Compatible with automated systems
  • Uniform external dimensions and tolerances
  • ANSI standard footprint
  • Alphanumeric well coding
  • Manufactured under DIN ISO 9001 guidelines
  • Sealable with adhesive films, capmat, and heat sealer
  • High temperature resistance
  • High chemical resistance
TRUSTED_SUSTAINABILITY

Specifications

Barcode Custom Barcoding Available
Format SBS Standard
Array 8 x 12
Color Natural
Disposable Limited-use
Product Type Microplate
Material Virgin Polypropylene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Square, Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) Sample Storage, Reagent Housing, Cultures
Green Features ACT Ecolabel (Silver)
No. per Pack 1 Ea.
Regulatory Status RUO
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.