Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Storage Vial
SDPSCI_ED

Catalog No. S15927 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S15927 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15927 Supplier United Scientific™ Supplier No. P60107

missing translation for 'validMainframeMsg'

DNase, RNase, and Pyrogen free.

  • Pack of 1,000 polypropylene storage vials with.
  • Ideal for storage of PCR reagents, enzymes and other diagnostics, biochemical reagents or samples.
  • Designed for use with samples from -80° to 121°C.
  • DNase, RNase, and Pyrogen free
  • Polypropylene vials feature a large white frosted labeling area, silk screen graduations, and a leak-proof HDPE cap
  • Autoclavable
  • Available in 2.0 mL or 5.0 mL

Specifications

Color Clear
Diameter (Metric) Outer 11.8 mm
Material Polypropylene
Pre-slit No
Sterility Non-sterile
Volume (Metric) 5 mL
For Use With (Application) PCR Reagents, Enzymes and Other Diagnostics, Biochemical Reagents
Closure Type Screw Cap
Quantity 1000/Pk.
Marking Spot Yes
Vial Type PCR Storage
Autoclavable Autoclavable
Height (Metric) 88 mm
Preservative None
Product Type Cryogenic Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.