Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Storage Vials
SDPSCI_ED

Catalog No. S04539 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S04539 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04539 Supplier United Scientific™ Supplier No. P60106
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use these Storage Vials for storage of PCR reagents, enzymes and other diagnostics and biochemical reagents/samples.

  • Made of polypropylene.
  • Each vial has an HDPE leakproof cap.
  • For use with samples from -80°C to +121°C.
  • DNase, RNase, and Pyrogen free.

Specifications

Color Clear
Diameter (Metric) Outer 11.8 mm
Material Polypropylene
Pre-slit No
Sterility Non-sterile
Volume (Metric) 2 mL
For Use With (Application) Samples from -80°C to +121°C
Closure Material HDPE
Closure Type Screw Cap
Quantity 1000 Pk.
Marking Spot Etched
Autoclavable Not Autoclavable
Height (Metric) 47.2 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.