Learn More
Saf T Pak Inc STP-340R - Category B / Exempt Insulated Shipping System (UN3373), Frozen w/ Reusable Secondary Pressure Vessel
SDP
Supplier:  Saf T Pak Inc STP340R
Encompass_Preferred
STP-340R Category B / Exempt Insulated Shipping System (UN3373), Frozen w/ Reusable Secondary Pressure Vessel, 2/Case Ship up to (6) STP-104 Reusable Secondary Pressure Vessels in one system. Ideal for long distance frozen shipments. Designed for use with Dry Ice or Saf-T-Temp products for thermal control. Outer Box - OD 18.5 X 15.25 X 15.5in., pre-printed with: Biological Substance Category B Marking, Exempt Human/Animal Speciment Marking, UN 3373 Marking, Orientation Arrows. Insulated Chest - ID 16 X 13 X 12.5in.
Catalog No. NC0404145