Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STRA8 Monoclonal Antibody (526CT11.1.5)
GREENER_CHOICE

Catalog No. PIMA524722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524722 Supplier Invitrogen™ Supplier No. MA524722
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STRA8 Monoclonal Antibody for Western Blot

Meiosis-inducer required for the transition into meiosis for both female and male germ cells. In female germ cells, required for premeiotic DNA replication and subsequent events in meiotic prophase. During spermatogenesis, next to its role in meiotic initiation, promotes (but is not required for) spermatogonial differentiation. Can associate with DNA (possibly in an indirect manner), and in vitro can activate DNA transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRA8
Applications Western Blot
Classification Monoclonal
Clone 526CT11.1.5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.09% sodium azide; pH 7.4
Gene STRA8
Gene Accession No. P70278, Q7Z7C7
Gene Alias RGD1562852; stimulated by retinoic acid 8; stimulated by retinoic acid 8 homolog; stimulated by retinoic acid gene 8; stimulated by retinoic acid gene 8 homolog; stimulated by retinoic acid gene 8 protein; Stimulated by retinoic acid gene 8 protein homolog; stra8
Gene Symbols STRA8
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 31-59 amino acids from human STRA8.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20899, 346673
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.