Learn More
Fine Science Tools Potts-Smith Forceps, Stainless Steel, Straight, 18 cm
SDP
Supplier:  Fine Science Tools 1102418
Encompass
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 1.25 x 1mm/ Alloy / Material: Stainless Steel/ Length: 18cm
Catalog No. NC9007346