Learn More
Excelta™ Straight Mosquito Clamp
Catalog No. 16050149 Shop All Excelta Corporation Products
Change view
Click to view available options
Tip Style:
Serrated
Smooth
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
16-050-149 Serrated
16-050-148 Smooth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-050-149 Supplier Excelta™ Supplier No. 33ASE
Used to clamp small blood vessels.
  • Locking mechanism allows the clamp to hold the vessel securely
  • Stainless Steel
  • Autoclavable
Specifications
Product Type Mosquito Clamp
Length (English) 5 in.
Length (Metric) 127 mm
Material Stainless Steel
Tip Style Serrated