Learn More
Krackeler Scientific Straight Needles
SDP
Catalog No. HP93010725N
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
HP93010725N 10 μL
HP51820875N 5 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. HP93010725N Supplier Krackeler Scientific Supplier No. 9293010725EA
Specifications
Needle Gauge 23 G
Length (English) Needle 1.65 in.
Product Type Straight Needle
Quantity 6 Pk.
Volume (Metric) 10 μL
Needle Point Style Cone
For Use With (Equipment) Autosampler Syringe
Connection Type Fixed