Learn More
Grainger Union Tee 1/2 in PK10
Supplier:  Grainger 16W614
Encompass_Preferred
The Parker 16W614 Union Tee is a composite union tee fitting made from engineered composite thermoplastic. It has a maximum pressure rating of 300 psi at 73°F. This union tee has socket end connections and is designed for use with Parker Parflex thermoplastic hose. The size is 1" NPT and it weighs 0.19 lbs. This composite fitting provides corrosion resistance and decreased weight versus metal fittings. The operating temperature range is -40°F to 150°F.
Catalog No. 50-276-3663
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.