Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Replacement Strap with Buckle for ArcGuard™ Suppression Blanket Storage Bag
SDP

Catalog No. 17001153 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Length (English):
4 ft.
6 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
17-001-153 4 ft.
17-001-154 6 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-001-153 Supplier National Safety Apparel™ Supplier No. ARCSTRAP4F
Add to Cart
Add to Cart

Aramid Fiber webbing strap for ArcGuard™ blanket.

Specifications

Width (English) 48 in.
Size Standard
Product Type Strap
Length (English) 4 ft.
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.