Learn More
CERILLO INC Stratus 4-Pack, Custom Wavelength
Supplier:  CERILLO INC 4PR2CUST
Encompass_Preferred
The Stratus 4-Pack, Custom Wavelength includes four Stratus plate readers at specified wavelengths, four USB cables, four power adapters, four 16 GB microSD cards, and 1 year of firmware upgrades. Custom Wavelegth (nm) must be specified upon order.
Catalog No. NC2603342
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.