Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ 3AA Helmet Lighting Kit

Catalog No. 19500562 Shop All Streamlight Products
Change view
Click to view available options
Helmet Lighting Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-500-562 Helmet Lighting Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-500-562 Supplier Streamlight™ Supplier No. 68270

Includes

3AA ProPolymer flashlight, three AA batteries, Streamlight helmet mount and rubber helmet strap. Helmet not included.

Attaches easily to any helmet without any tools

Waterproof, nonconductive, and corrosion proof.

  • Long-range brilliance and smooth uniform pattern
  • Maximum light output throughout battery life
  • Yellow, impact-resistant body and lens cover
  • High-strength rubber flashlight strap for helmets offers quick and easy, no-tools-required method to attach flashlight to almost any helmet
  • Lightweight aluminum, flashlight mount provides a low-profile fit
  • Light source: one C4™ LED; 50,000 hr. LED life
  • Light output: 120 lumens, (up to 14,000 candela)
  • Runtime: 6.5 hr.
  • Lens: unbreakable polycarbonate lens with scratch-resistant coating
  • Switch: rubber-booted, tail-end, push button switch for one-handed operation

Order Info

L x W x D: 7 x 1.5 x 1.05 in. (17.78 x 3.81 x 2.67cm)

Certifications

UL listed Class I Division I hazardous locations

Specifications

Light Source C4 LED
Weight (English) 7.52 oz.
Weight (Metric) 0.21 kg
Battery Type 4 AA Alkaline
Controller Type Rubber booted, tail-end push button-switch
Diameter (English) Body 1.48 in.
Diameter (Metric) Body 3.76 cm
Dimensions (L x W) 17.78 x 3.43 cm (7 x 1.35 in.)
For Use With (Application) Attaches easily to any helmet without any tools
Includes 4 AA ProPolymer™ Lux Div 2, black jack helmet mount, rubber helmet strap
Length (English) 7 in.
Length (Metric) 17.78 cm
Lens Material Polycarbonate
Material Polymer Resin
Width (English) 1.35 in.
Width (Metric) 3.43 cm
Product Type Helmet Lighting Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.