Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ 3C ProPolymer™ Lux Flashlight

Catalog No. 19819889 Shop All Streamlight Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-889 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-889 Supplier Streamlight™ Supplier No. 33702

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Features a deep parabolic reflector that produces long range brilliance and a smooth uniform light pattern

  • 10,000 Hour, 1 watt super high-flux lux LED
  • Up to 3500 candela, 44 lumens
  • Powered by three C alkaline batteries, not included
  • Runs up to four hours at full intensity; up to 15 hrs to 50 percent output intensity
  • Rubber booted, tail-end push button-switch
  • Momentary and constant on
  • Spring-loaded clip
  • Water resistant, O-ring sealed (not dive rated)
  • Corrosion proof and non-conductive
  • Unbreakable polycarbonate lens with scratch resistant coating
  • Shock resistant

Order Info

Length: 9.3 in. (23.6cm). Weight (with batteries): 14.6 oz. (414g).

Certifications

UL listed for Class I, Division 1; Class II, Division 1; and Class III hazardous locations; Exia Temperature Code T4.

Specifications

Light Source LED
Weight (English) 15 oz.
Weight (Metric) 0.42 kg
Battery Life 10,000 hr.
Battery Type C Alkaline
Certifications/Compliance UL Class I, Division 1, Class II, Division 1 and Class III, T4
Color Black
Controller Type Rubber booted, tail-end push button-switch
Includes White LED
Length (English) 9.3 in.
Length (Metric) 23.6 cm
Lens Material Polycarbonate
Lumen 44 lm
Material Polymer
Product Type LED Flashlight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.