Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ Replacement Parts For LiteBox™ FireBox™ Rechargeable Lanterns

Catalog No. 19028323 Shop All Streamlight Products
Change view
Click to view available options
Product Type:
Flashlight Kit
Flood Lamp Assembly
Housing Flashlight
PCB Standard Assembly Kit
Spot Lamp Assembly
Streamlight Battery
Streamlight Bulb
For Use With (Equipment):
Litebox, FireBox Lanterns
Litebox, FireBox Rechargeable Lanterns
PAR 36 Lamp Assembly
SL-40 Flashlights
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
19-028323 Housing Flashlight SL-40 Flashlights
19-822-939 Flashlight Kit PAR 36 Lamp Assembly
19-823-887 PCB Standard Assembly Kit Litebox, FireBox Rechargeable Lanterns
19-819-834 Streamlight Battery Litebox, FireBox Lanterns
19-822-941 Spot Lamp Assembly Litebox, FireBox Rechargeable Lanterns
19-819-831 Streamlight Bulb Litebox, FireBox Lanterns
19-819-832 Flood Lamp Assembly Litebox, FireBox Lanterns
19-819-833 Spot Lamp Assembly Litebox, FireBox Lanterns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-028323 Supplier Streamlight™ Supplier No. 400255

Specifications

Product Type Housing Flashlight
Color Orange
For Use With (Application) Fire, Emergency and Rescue
For Use With (Equipment) SL-40 Flashlights
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.