Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ Vulcan™ Rechargeable Lantern Systems

Catalog No. 19041388 Shop All Streamlight Products
Change view
Click to view available options
Color:
Orange
Yellow
Includes:
Shoulder Strap, AC/DC Charger
Shoulder Strap, Charging/Mounting Rack, AC/DC Charger
Ultrabright Taillight LEDs
Lumen:
150 lm
157 lm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes Lumen
19-041-388 Yellow Shoulder Strap, AC/DC Charger 157 lm
19-121-807 Orange Ultrabright Taillight LEDs 150 lm
19-186-170 Yellow Shoulder Strap, Charging/Mounting Rack, AC/DC Charger 157 lm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-041-388 Supplier Streamlight™ Supplier No. 44200

Powerful lantern comes in fire service version with LED tail lights

System Components

  • Rechargeable flashlight with cushioned grip handle and a high-impact, waterproof, ABS thermoplastic housing
  • 6V rechargeable battery
  • 120VAC and 12VDC power cords
  • Halogen bi-pin bulb
  • 80,000 candela
  • Charging/mounting rack, which meets requirements of NFPA 1901-12-1.7 (1999) mounted in any position
  • Shoulder strap

Fire Vulcan™ Option

  • Extremely powerful (65,000 candlepower) fire service flashlight features two ultrabright blue tail light LEDs that allow you to see others in smoke-filled rooms
  • With quick-release strap and glove-friendly, three-position ON/OFF toggle switch

Order Info

Maximum candlepower with supplied 8w spot: up to 80,000. Run time: 3.25 hr. on regular Vulcan; Fire Vulcan option provides 3 hours. Weight: 3.3 lb. (1.5kg). Length: 7.5 in. (19cm).

Certifications

USTC approved for Class l, ll, lll, Division 2. Most models meet applicable European Community Directives, including ATEX Zone 2

Compliance

Rack meets requirements of NFPA 1901-12-1.7 mounted in any position.

Warranty and Services

Limited lifetime

Specifications

Light Source Halogen
Lighting Life 3.25 hr.
Wattage 8 W
Weight (English) 3.3 lb.
Weight (Metric) 1.5 kg
Battery Type 4.5 AH Lead Acid
Certifications/Compliance Class l, ll, Ill, Division 2, NFPA 1901-12-1.7, Class I, II, Division 2, Class III
Color Yellow
Description Dual filament
For Use With (Application) Ideal for Field Maintenance, Inspection of Construction Sites and Remote Equipment
Height (English) 6.5 in.
Height (Metric) 16.5 cm
Includes Shoulder Strap, AC/DC Charger
Length (English) 7.5 in.
Length (Metric) 17.05 cm
Lumen 157 lm
Material ABS Thermoplastic
Width (English) 5 in.
Width (Metric) 12.7 cm
Product Type Rechargeable Lantern
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.