Learn More
Description
System Components
- Rechargeable flashlight with cushioned grip handle and a high-impact, waterproof, ABS thermoplastic housing
- 6V rechargeable battery
- 120VAC and 12VDC power cords
- Halogen bi-pin bulb
- 80,000 candela
- Charging/mounting rack, which meets requirements of NFPA 1901-12-1.7 (1999) mounted in any position
- Shoulder strap
Fire Vulcan™ Option
- Extremely powerful (65,000 candlepower) fire service flashlight features two ultrabright blue tail light LEDs that allow you to see others in smoke-filled rooms
- With quick-release strap and glove-friendly, three-position ON/OFF toggle switch
Order Info
Maximum candlepower with supplied 8w spot: up to 80,000. Run time: 3.25 hr. on regular Vulcan; Fire Vulcan option provides 3 hours. Weight: 3.3 lb. (1.5kg). Length: 7.5 in. (19cm).
Certifications
USTC approved for Class l, ll, lll, Division 2. Most models meet applicable European Community Directives, including ATEX Zone 2
Compliance
Rack meets requirements of NFPA 1901-12-1.7 mounted in any position.
Warranty and Services
Limited lifetime
Specifications
Specifications
| Light Source | Halogen |
| Lighting Life | 3.25 hr. |
| Weight (English) | 3.3 lb. |
| Weight (Metric) | 1.5 kg |
| Battery Type | 4.5 AH Lead Acid |
| Certifications/Compliance | Class l, ll, Ill, Division 2, NFPA 1901-12-1.7, Class I, II, Division 2, Class III |
| Color | Orange |
| Height (English) | 6.5 in. |
| Height (Metric) | 16.5 cm |
| Includes | Ultrabright Taillight LEDs |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.