Learn More
Description
- Assayed for 30 parameters, including T-lymphocytes, B-lymphocytes and NK cells.
- Contains a CD34+ population and is available in two clinically relevant CD4+ levels.
- Compatible with BD Biosciences and Beckman Coulter flow cytometry systems.
- 30-day open-vial stability, 90-day closed-vial stability.
- Manufacturer established assay ranges – no need to verify range per CAP requirements.
- Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | 2°C to 10°C |
| Format | Cap-pierceable Plastic Vial |
| For Use With (Equipment) | Beckman Coulter™ flow cytometry system |
| Type | CD-Chex™ plus Immunophenotyping Control |
| Includes | 2 x 3 mL Low, 2 x 3 mL Normal |
| Intended Use | Immunophenotyping including RBC lysis, Antibody staining, instrument performance and operator technique |
| Level | CD4 Low, Normal |
| Quantity | 4 x 3 mL |
| Source | Human Peripheral Blood Leukocytes and Erythrocytes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

