Learn More
Description
- Assayed for the 5-part WBC differential.
- Contains human white blood cells and CPPD crystals.
- 30-day open-vial stability, 6-month closed-vial stability.
- Validates the entire cell qualification and quantification procedure.
- Long shelf life reduces the number of shipments and associated costs.
- Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2°C to 10°C |
| For Use With (Equipment) | A hemacytometer or WBC differential counts using a cytocentrifuge smear |
| Type | Cell Body Fluid Control |
| Intended Use | Body fluid procedural control for RBC and WBC counts, crystal identification and WBC differentiation |
| Level | 2 |
| Quantity | 6 x 2 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

