Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck CVA™ Calibration Verification Assessment/Linearity Kit
GSA_VASDP

Catalog No. 11716366 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
5 x 3 mL
13 x 3 mL
16 x 3 mL
18 x 3 mL
Intended Use:
CVA for RVC, HgB
CVA for WBC, Platelets, HgB
For Use With (Equipment):
ADVIA 120 and LH750
ADVIA 60 Hematology Analyzer
CELL DYN 3200
HemoCue Hematology Analyzer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Intended Use For Use With (Equipment)
11-716-366 16 x 3 mL CVA for WBC, Platelets, HgB CELL DYN 3200
11-716-320 5 x 3 mL CVA for RVC, HgB HemoCue Hematology Analyzer
11-716-360 18 x 3 mL CVA for WBC, Platelets, HgB ADVIA 120 and LH750
23-285-733 13 x 3 mL CVA for WBC, Platelets, HgB ADVIA 60 Hematology Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11-716-366 Supplier Streck Supplier No. 291718
This item is not returnable. View return policy

CVA (Calibration Verification Assessment) and CVA for CELL-DYN are assayed linearity control kits that test the upper and lower limits of three- and five-part differential hematology analyzers to determine their patient reportable range and linear performance.

  • Provides assay values for the most popular three- and five-part hematology instruments.
  • Customized kits with WBC, RBC, Hgb and platelet ranges for each instrument model.
  • Plastic vials with pierceable caps for analyzer autosampling.
  • 5-day open-vial stability, 120-day closed-vial stability.
  • Tests the extreme upper and lower limits of an instrument.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

For Use With (Equipment) CELL DYN 3200
Type Linearity Control Kit
Includes WBC, platelet and RBC or Hemoglobin vials
Intended Use CVA for WBC, Platelets, HgB
Level 5
Quantity 16 x 3 mL
Stability 5 Days Open, 120 Days Closed Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.