Description
Includes
Six vials of receiver solution and one or two vials of pipetting solution
- Easy-to-use kits are shipped to the lab.
- NIST-traceable.
- Compatible with any pipette brand.
- Rapid turnaround.
- Pipettes do not have to leave the lab.
- Users receive an email version of a regulatory-compliant report.
Order Info
Order one PVS kit per pipet to be tested
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Product Type | Maintenance Kit |
| For Use With (Application) | To perform simple titrations, analysis and calculation of the results |
| Volume (Metric) | 10 to 200 μL |
| Includes | Six vials of receiver solution and one or two vials of pipetting solution. |
| Quantity | 1 Ea. |