Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Sickledex™ Solubility Kit
SDP

Catalog No. 11716450 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
100 Tests
12 Tests
50 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-716-450 100 Tests
23-285-555 12 Tests
23-285-625 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-716-450 Supplier Streck Supplier No. 217657
Add to Cart
Add to Cart

Includes

Each test kit contains two bottles of phosphate buffer with Saponin, two buffer dispensing caps and two vials of Sodium Hydrosulfite reagent powder.

Requires

Each test requires 2 mL of reconstituted solubility buffer and 20μL of anticoagulated whole blood or 20μL of sickle cell hemoglobin control.

SICKLEDEX is a qualitative solubility test that can detect the presence of sickling hemoglobin in human blood samples within 6 minutes. With this kit, your laboratory is supplied with items necessary to perform easy and efficient sickle solubility testing, including the SICKLEDEX solubility test kit, Sickle-Chex™ whole blood control and a convenient test tube rack.

  • Test results available in 6 minutes.
  • Requires only 20 µL of patient or control sample.
  • 45-day open-vial stability, 12-month closed-vial stability.
  • Buffer packaging reduces waste and maximizes open-vial stability.
  • No pipetting necessary.
  • Sickle-Chex whole blood control can be used like a patient sample.

Order Info

Additional test tube racks are available separately, Cat. No. 11-716-452. Additional test tubes, Cat. No. NC1459622.

Specifications

Content And Storage 2°C to 10°C
Type Sickle Cell Solubility Test Kit
Includes 2 Bottles of Phosphate Buffer with Saponin, 2 Buffer Dispensing Caps, 2 Vials of Sodium Hydrosulfite Reagent Powder
Quantity 100 Tests
Test Time 6 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.