Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Strep B Carrot Broth™ OneStep
SDP

Catalog No. 23001733 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Fill Volume:
4 mL
6 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
23-001-733 4 mL
23-001-746 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-001-733 Supplier Hardy Diagnostics™ Supplier No. Z40
Add to Cart
Add to Cart
This item is not returnable. View return policy

A selective and differential medium intended for the detection of Group B Streptococcus (GBS) from anovaginal specimens collected from pregnant women.

  • The color change reaction from white to orange is representative of a positive result for the presence of hemolytic GBS
  • Our one-step test is an improvement over conventional methods by increasing sensitivity, decreasing turn-around time, and lowering overall cost
  • All components are now included in the color change test, so the use of additional tiles is no longer necessary
  • Accurate results for positives, there is no need for further sub-culturing or testing
  • Can be used with liquid and gel-based transport systems

Specifications

Appearance Hazy to Cloudy, Colorless Liquid
Broth Type Carrot Broth
Certifications/Compliance ISO 13485
Color Orange
Content And Storage 2°C to 8°C
Additive Selective Agents
Detects Group B Streptococcus
Format Liquid
Manufacturing Origin United States
Media Type Broth
Product Type Broth Tube
Test Type Microbial Identification
Fill Volume 4 mL
Formulation Peptone - 25.0 gm, Starch - 10.0 gm, Selective Agents - 38.8 mg, Morpholinepropanesulfonic Acid (MOPS) - 11.0 gm, Disodium Phosphate - 8.5 gm, Dextrose - 2.5 gm, Sodium Pyruvate - 1.0 gm, Magnesium Sulfate - 0.2 gm
For Use With (Application) Detection of Group B Streptococcus (GBS)
Incubation Conditions Aerobically at 35°C
Incubation Time 16 to 24 hr.
Material Glass
No. per Pack 20
Packaging Type Box
Quantity 20 Ea.
Transfer Volume 30 μL
Volume (Metric) Specimen 30 μL
Dimensions (Dia. x H) 13 x 100 mm
Height (Metric) 100 mm
Diameter (Metric) 13 mm
Species Streptococcus species
Manufacturing Quality ISO 13485
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.