Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Strep-Diluent
SDP

Catalog No. 48283012 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
7.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-283-012 7.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-283-012 Supplier R&D Systems™ Supplier No. 48283012

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ Strep-Diluent is an optimized blocking reagent specifically developed to dilute Strep-HRP and block unspecific binding in the TACS XL™ line of kits.

  • Uncontrolled product according to WHMIS classification criteria
  • All of the ingredients in this product are listed on the United States TSCA (Toxic Substances Control Act) inventory, the Domestic Substances List (DSL) and/or the Non-Domestic Substances List (NDSL)

Specifications

Quantity 7.5 mL
Content And Storage Store the unopened product at 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.