Learn More
IBA LifeSciences Strep-Tactin conjugate APC; 50 µl
Supplier:  IBA LifeSciences 65010001
Encompass_Preferred
Strep-Tactin conjugated to either R-Phycoerythrin (PE) or Allophycocyanin (APC) is suitable for staining or isolation of antigen-specific cytotoxic T cells (MHC I Streptamer) or surface marker-specific cells (Fab-TACS/Nano-TACS) based on our Strep-tag technology. To target specific cells, MHC I complexes, low affinity Fab fragments or nanobodies fused to a Twin-Strep-tag (MHC I-Streps, Fab-Streps or Nano-Streps) are required in addition to fluorescent Strep-Tactin. Strep-Tactin multimerizes MHC I-Streps or Fab-/Nano-Streps, ensuring stable binding to cells. The resulting fluorescent complexes can be used to stain cells that are specific for the respective peptide-loaded MHC I molecule or for a selected surface marker. The addition of biotin reverses the multimerization and causes the dissociation of MHC I-Streps or Fab-/Nano-Streps from the cell surface. The complete removal of all labeling reagents preserves the cells’ full biological activity.
Catalog No. 50-210-0167
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.