Learn More
IBA LifeSciences StrepMAB-Immo DY-549 conjugate , 50 µg
Supplier:  IBA LifeSciences 21567050
Encompass_Preferred
StrepMAB-Immo DY-549 is a murine monoclonal antibody conjugated to a fluorescent dye. It has a high affinity to Strep-tag and Twin-Strep-tag fusion proteins, and can be used for detection in ELISA, FACS, immunohistochemistry and immunocytochemistry.For proteins carrying the Strep-tag with an N-terminal Serine-Alanine linker (protein-SA-WSHPQFEK or SA-WSHPQFEK-protein), the binding affinity is in the pM range. Proteins with other N-terminal sequences bind with a reduced affinity. The Twin-Strep-tag includes an N-terminal SA-linker (WSHPQFEK-GGGS-GGGS-GGSA-WSHPQFEK).The absorption/emission maxima of the fluorescent dye is 550-560/575 nm
Catalog No. NC2604675
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.