Learn More
Description
- Highly pure Strep II-tagged recombinant proteins eluted in concentrated form and small volumes
- Physiological conditions and mild elution preserve target protein activity
- Fast and easy regeneration with 0.5M NaOH
- Compatible with a wide range of reducing agents, detergents, denaturants and other additives
Specifications
Specifications
| Quantity | 50 mL |
| Flow Rate | <300cm/h |
| Ligand Type | StrepTactin |
| Content And Storage | 2 to 8°C, 20% Ethanol |
| Matrix | Affinity Resin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.