Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptavidin, lyophilized powder

Catalog No. 434301
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-430-1 1 mg
43-430-2 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-430-1 Supplier Invitrogen™ Supplier No. 434301
Add to Cart
Edge
Add to Cart

A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.

  • A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.
  • Binding activity of streptavidin is determined to be >11 units per mg protein
  • One unit of binding activity is defined as that amount of protein which binds 1 μg of biotin (Green, N.M., Biochem. J., 89:599,1963)
  • Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4
  • Reconstitute with 1 mL of distilled or deionized water
  • Store at 4°C
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Unconjugated
Quantity 1 mg
Content And Storage Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4. Reconstitute with 1 ml of distilled or deionized water. Store at 4°C.
Shipping Condition Wet Ice
Form Lyophilized
Product Type Streptavidin Conjugate (fluorescent)
Are there any His-tags on Streptavidin (Cat. No. 434302)?

This Streptavidin is a native protein from Streptomyces avidinii, therefore there are no His-tags on it.

How many biotin binding-sites are there per streptavidin molecule?

Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.

Is Streptavidin (Cat. No. S888, 434302, and 434301) RNAse-free?

Although we utilize manufacturing methods that minimize the introduction of any contaminants such as RNases, we do not process this product to specifically remove or inactivate RNases and do not test for RNase activity.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.