Learn More
ACROBiosystems Streptavidin Coated Plates, Clear, 96-Well, Clear Frame (For Serological Testing) 5 plate
Supplier:  ACROBiosystems SP155PLATES
Encompass_Preferred
The plate could not be separated, and the frame is clear. Based on the high binding affinity between biotin-streptavidin systems, our best seller SP-15 Streptavidin Coated Plates (For Serological Testing) are recommended to use together with biotinylated proteins to get lower non-specific binding results in a shorter time and be easy to watch color changes.
Catalog No. 50-233-1681
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.