Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ Streptavidin - Agarose

Catalog No. 5075987
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-759-87 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-759-87 Supplier Molecular Probes™ Supplier No. 434341
Add to Cart
Edge
Add to Cart

Streptavidin - Agarose

Reactivity: SA-Seph 4B reacts with biotin.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg Streptavidin/mL Sepharose Gel
Quantity 2 mL
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Wet Ice
Form Liquid Suspension
Product Type Streptavidin Conjugate (Fluorescent)
How many biotin binding-sites are there per streptavidin molecule?

Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.