Learn More
XpressBio Xpress Capture Streptavidin Coated ELISA Plates, 96 Clear Wells (5-pack)
SDP
Supplier:  XpressBio EX8000C96
Encompass_Preferred
Capture coated Streptavidin ELISA plates may be used with biotinylated peptides, antigens, antibodies, DNA, and oligonucleotides for various uses such as immunoassays and DNA/RNA quantitation.
Catalog No. 50-171-7652
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.