Learn More
R&D Systems™ Streptavidin-Fluorescein
SDP
Catalog No. 48003014 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
30 μL
Catalog No. Quantity
48003014 30 μL
1 options
This item is not returnable. View return policy
Catalog No. 48003014 Supplier R&D Systems™ Supplier No. 48003014
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
For use with apoptosis detection reagents and kits
Specifications
Quantity 30 μL
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Type Streptavidin-Fluorescein
Product Line R&D Systems