Learn More
AdipoGen Streptavidin HRP
SDP
Supplier:  AdipoGen AG35B0014EC050
Encompass_Preferred
Recombinant protein. Recombinant streptavidin from Streptomyces avidinii. Binds to any biotinylated reagents. Endotoxin . Lyophilized. Contains PBS. Streptavidin is a non-glycosylated protein isolated from Streptomyces avidinii. It forms a homotetramer and strong non-covalent specific complex with biotin one molecule of biotin per subunit of streptavidin. Streptavidin HRP is useful as a second step reagent for indirect enzymatic labeling, in conjunction with biotinylated primary antibodies. It is often used in ELISA, IHC or Western blotting applications together with relevant substrate systems.
Catalog No. 50-256-3077
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.