Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Streptavidin PerCP-Cyanine5.5 Conjugate

Catalog No. 5015835
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-158-35 25 μg
50-158-36 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Contains Sodium azide
Catalog No. 50-158-35 Supplier Invitrogen™ Supplier No. 45431780
Add to Cart
Add to Cart
Contains Sodium azide

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Blue Laser

Emit

695 nm

Excite

488 nm

Reported Application

Flow Cytometric Analysis

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.2 mg/mL
Conjugate PerCP-Cy5.5
Quantity 25 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye PerCP-Cy5.5
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.