Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Streptavidin Phycoerythrin, Recombinant Protein, Flow Cytometry, 500 μL
Description
- Source: Streptomyces avidinii expressed in E. coli
- Tested Applications: Flow Cyt
- Form: Liquid
- Purification technique: Size-exclusion chromatography
- Formulation: pH 7.2, 0.09% Sodium azide, 2.9% Sodium chloride, and 0.12% Sodium dihydrogen phosphate
- Concentration: 0.2 mg/mL
- Storage Conditions: Store at +4°C
Specifications
Specifications
| Target Molecule | Biotin |
| Shipping Condition | Blue Ice |
| For Use With (Equipment) | Flow Cytometer |
| Label or Dye | Phycoerythrin |
| Form | Liquid |
| Buffer | 2.9% sodium chloride, 0.12% sodium dihydrogen phosphate, 0.09% sodium azide, pH7.2 |
| Concentration | 0.2 mg/mL |
| Content And Storage | Store at 4°C. |
| Detection Method | Fluorescence |
| Formulation | pH 7.2, 0.09% Sodium azide, 2.9% Sodium chloride, 0.12% Sodium dihydrogen phosphate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?